Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152230.4 | NP_689416.1 | 3 | 646 | ||
XM_011539565.2 | XP_011537867.1 | 3 | 687 | |||
XM_017016013.1 | XP_016871502.1 | 3 | 313 | |||
GenBank mRNA | AF432853.1 | - | 3 | 323 | ||
BC065709.1 | - | 3 | 650 | 6654071 |