Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK38L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015000.3 | NP_055815.1 | 14 | 1834 | ||
XM_005253342.3 | XP_005253399.1 | 13 | 1713 | |||
XM_006719058.3 | XP_006719121.1 | 15 | 1815 | |||
XM_006719059.3 | XP_006719122.1 | 15 | 1794 | |||
XM_011520613.2 | XP_011518915.1 | 13 | 1528 | |||
XM_017019036.1 | XP_016874525.1 | 14 | 1790 | |||
GenBank mRNA | AB023182.2 | - | 14 | 1919 | ||
AK304862.1 | - | 8 | 1130 | |||
BC028603.1 | - | 14 | 1808 | 4823429 |