Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321781.1 | NP_001308710.1 | 3 | 555 | ||
NM_001321782.1 | NP_001308711.1 | 3 | 548 | |||
NM_152516.3 | NP_689729.1 | 3 | 652 | |||
XM_011532559.2 | XP_011530861.1 | 3 | 603 | |||
XM_017003412.1 | XP_016858901.1 | 3 | 460 | |||
GenBank mRNA | AB178811.1 | - | 3 | 636 | ||
AK223607.1 | - | 3 | 641 | |||
BC009266.2 | - | 3 | 605 | |||
BC022046.1 | - | 3 | 624 | 4046433 | ||
BG182961.1 | - | 1 | 158 | |||
BG183486.1 | - | 1 | 158 | |||
BG215225.1 | - | 1 | 158 | |||
DT217730.1 | - | 3 | 554 |