Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152540.3 | NP_689753.2 | 3 | 1178 | ||
XM_011534375.2 | XP_011532677.1 | 3 | 1214 | |||
XM_011534376.2 | XP_011532678.1 | 3 | 1214 | |||
XM_011534378.2 | XP_011532680.1 | 3 | 1214 | |||
XM_017007786.1 | XP_016863275.1 | 3 | 1214 | |||
XM_017007787.1 | XP_016863276.1 | 3 | 1214 | |||
GenBank mRNA | AB074276.1 | - | 3 | 1151 | ||
AK096833.1 | - | 3 | 1164 | |||
AY299407.1 | - | 3 | 1178 | |||
BC012458.1 | - | 3 | 944 | |||
BC021137.1 | - | 3 | 1135 | 4866354 | ||
BC032453.1 | - | 3 | 1176 | 5176029 |