Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other USP54 SIRNA Assays ›
Gene Symbol:
USP54
Gene Name:
ubiquitin specific peptidase 54
Gene Aliases:
C10orf29, bA137L10.3, bA137L10.4
Chromosome Location:
Chr.10: 73497538 - 73626844 on Build GRCh38
UniGene ID:
Hs.657355
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
148141
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: USP54
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320437.1
NP_001307366.1
22
4486
NM_152586.3
NP_689799.3
22
4599
NR_135249.1
22
4845
NR_135250.1
18
3966
XM_005269582.2
XP_005269639.1
20
7536
XM_011539368.2
XP_011537670.1
22
7743
XM_017015759.1
XP_016871248.1
24
7172
XM_017015760.1
XP_016871249.1
24
6074
XM_017015761.1
XP_016871250.1
24
6912
XM_017015762.1
XP_016871251.1
24
6082
XM_017015763.1
XP_016871252.1
24
5376
XM_017015764.1
XP_016871253.1
24
5439
XM_017015765.1
XP_016871254.1
25
5692
XM_017015766.1
XP_016871255.1
24
7298
XM_017015767.1
XP_016871256.1
24
6957
XM_017015768.1
XP_016871257.1
24
5128
XM_017015769.1
XP_016871258.1
24
4736
XM_017015770.1
XP_016871259.1
24
5397
XM_017015771.1
XP_016871260.1
23
7914
XM_017015772.1
XP_016871261.1
23
7905
XM_017015773.1
XP_016871262.1
22
7854
XM_017015774.1
XP_016871263.1
23
6840
XM_017015775.1
XP_016871264.1
24
6718
XM_017015776.1
XP_016871265.1
23
7223
XM_017015777.1
XP_016871266.1
23
6882
XM_017015778.1
XP_016871267.1
22
7800
XM_017015779.1
XP_016871268.1
22
7779
XM_017015780.1
XP_016871269.1
22
7764
XM_017015781.1
XP_016871270.1
22
7749
XM_017015782.1
XP_016871271.1
23
5500
XM_017015783.1
XP_016871272.1
23
6547
XM_017015784.1
XP_016871273.1
22
4493
XM_017015785.1
XP_016871274.1
21
7659
XM_017015786.1
XP_016871275.1
22
4990
GenBank mRNA
AK094637.1
-
4
1552
AK123711.1
-
4
573
AK126497.1
-
10
2954
AK127213.1
-
8
2393
AK127267.1
-
18
3966
AK303633.1
-
6
2051
AL832102.1
-
4
1498
BC026037.1
-
5
1447
BC064967.1
-
4
1494
BC130633.1
-
6
2042
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
protein deubiquitination
protein binding
thiol-dependent ubiquitinyl hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline