Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008707.1 | NP_001008707.1 | 23 | 3796 | ||
NM_004434.2 | NP_004425.2 | 22 | 3739 | |||
XM_005267397.1 | XP_005267454.1 | 23 | 3866 | |||
XM_005267398.3 | XP_005267455.1 | 23 | 3799 | |||
XM_005267399.4 | XP_005267456.2 | 23 | 3838 | |||
XM_005267400.4 | XP_005267457.1 | 22 | 3765 | |||
XM_011536540.2 | XP_011534842.1 | 22 | 3812 | |||
XM_011536541.1 | XP_011534843.1 | 23 | 3879 | |||
XM_011536542.2 | XP_011534844.1 | 22 | 3706 | |||
XM_017021074.1 | XP_016876563.1 | 21 | 3677 | |||
GenBank mRNA | AF035276.1 | - | 1 | 403 | ||
AK126980.1 | - | 24 | 3752 | |||
BC033043.1 | - | 22 | 3739 | 4821202 | ||
U97018.1 | - | 22 | 3745 |