Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304963.1 | NP_001291892.1 | 2 | 771 | ||
NM_153183.3 | NP_694853.1 | 3 | 834 | |||
GenBank mRNA | AF469196.1 | - | 3 | 836 | ||
AK291952.1 | - | 2 | 751 | |||
BC049383.1 | - | 2 | 692 | 5725355 | ||
BC050700.1 | - | 2 | 750 | 5723622 | ||
BX449265.2 | - | 2 | 771 | |||
CN399689.1 | - | 2 | 440 |