Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080551.2 | NP_001074020.2 | 7 | 1049 | ||
NM_173521.4 | NP_775792.4 | 9 | 1056 | |||
NR_109816.1 | 11 | 1258 | ||||
XM_006716974.2 | XP_006717037.1 | 11 | 1242 | |||
XM_006716975.2 | XP_006717038.1 | 11 | 1242 | |||
XM_011518302.2 | XP_011516604.1 | 9 | 3051 | |||
XM_011518303.1 | XP_011516605.1 | 9 | 1023 | |||
XM_011518306.1 | XP_011516608.1 | 6 | 643 | |||
XM_011518309.1 | XP_011516611.1 | 11 | 1242 | |||
XM_017014340.1 | XP_016869829.1 | 9 | 1860 | |||
XM_017014341.1 | XP_016869830.1 | 11 | 1242 | |||
GenBank mRNA | AK057341.1 | - | 11 | 1193 | ||
AK126055.1 | - | 7 | 1049 | |||
AL833535.1 | - | 9 | 1050 | |||
BC112357.1 | - | 11 | 1186 | |||
BC144680.1 | - | 11 | 1182 |