Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080551.2 | NP_001074020.2 | 8 | 1143 | ||
NM_173521.4 | NP_775792.4 | 10 | 1150 | |||
NR_109816.1 | 12 | 1352 | ||||
XM_006716974.2 | XP_006717037.1 | 12 | 1336 | |||
XM_006716975.2 | XP_006717038.1 | 12 | 1336 | |||
XM_011518302.2 | XP_011516604.1 | 10 | 3145 | |||
XM_011518303.1 | XP_011516605.1 | 10 | 1117 | |||
XM_011518306.1 | XP_011516608.1 | 7 | 737 | |||
XM_011518309.1 | XP_011516611.1 | 12 | 1336 | |||
XM_017014340.1 | XP_016869829.1 | 10 | 1954 | |||
XM_017014341.1 | XP_016869830.1 | 12 | 1336 | |||
GenBank mRNA | AK057341.1 | - | 12 | 1287 | ||
AK126055.1 | - | 8 | 1143 | |||
AL833535.1 | - | 10 | 1144 | |||
BC112357.1 | - | 12 | 1280 | |||
BC144680.1 | - | 12 | 1276 |