Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020945.1 | NP_065996.1 | 7 | 951 | ||
XM_005270004.3 | XP_005270061.1 | 7 | 1066 | |||
XM_011539986.2 | XP_011538288.2 | 7 | 1069 | |||
XM_011539987.2 | XP_011538289.2 | 7 | 1072 | |||
XM_011539988.2 | XP_011538290.1 | 8 | 1150 | |||
XM_011539990.2 | XP_011538292.1 | 7 | 1097 | |||
XM_011539991.2 | XP_011538293.1 | 7 | 1101 | |||
XM_011539992.2 | XP_011538294.1 | 7 | 1101 | |||
XM_017016463.1 | XP_016871952.1 | 8 | 993 | |||
XM_017016465.1 | XP_016871954.1 | 7 | 1078 | |||
GenBank mRNA | AK127650.1 | - | 7 | 960 | ||
BC034937.1 | - | 7 | 1017 |