Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243313.1 | NP_001230242.1 | 4 | 539 | ||
NM_015690.4 | NP_056505.2 | 4 | 539 | |||
XM_005246464.1 | XP_005246521.1 | 4 | 427 | |||
XM_011510959.1 | XP_011509261.1 | 4 | 539 | |||
XM_011510960.1 | XP_011509262.1 | 4 | 480 | |||
XM_017003804.1 | XP_016859293.1 | 4 | 427 | |||
XM_017003805.1 | XP_016859294.1 | 3 | 207 | |||
GenBank mRNA | AB033104.1 | - | 4 | 496 | ||
AF200815.1 | - | 3 | 260 | |||
BC026158.2 | - | 4 | 479 | 4839214 |