Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244959.1 | NP_001231888.1 | 4 | 514 | ||
NM_001244960.1 | NP_001231889.1 | 4 | 290 | |||
NM_174938.5 | NP_777598.3 | 4 | 514 | |||
XM_017014588.1 | XP_016870077.1 | 4 | 204 | |||
GenBank mRNA | AK094281.1 | - | 4 | 215 | ||
AK223597.1 | - | 4 | 441 | |||
AK297722.1 | - | 4 | 432 | |||
AY137774.1 | - | 4 | 298 | |||
EF560742.1 | - | 4 | 365 |