Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SESTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178123.4 | NP_835224.3 | 4 | 342 | ||
XM_011512139.2 | XP_011510441.1 | 4 | 370 | |||
XM_011512141.2 | XP_011510443.1 | 2 | 47 | |||
XM_017005250.1 | XP_016860739.1 | 4 | 319 | |||
XM_017005251.1 | XP_016860740.1 | 4 | 442 | |||
GenBank mRNA | AK096232.1 | - | 4 | 320 | ||
AY209190.1 | - | 4 | 239 | |||
BC047578.1 | - | 4 | 491 | 4815542 |