Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183376.2 | NP_899232.2 | 8 | 2516 | ||
GenBank mRNA | AK026436.1 | - | 3 | 1493 | ||
BC028704.2 | - | 8 | 2511 | 4838787 | ||
BC070100.1 | - | 8 | 2501 | 30345612 |