Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323977.1 | NP_001310906.1 | 5 | 734 | ||
NM_001323978.1 | NP_001310907.1 | 6 | 1162 | |||
NM_001323979.1 | NP_001310908.1 | 5 | 849 | |||
NM_001323980.1 | NP_001310909.1 | 5 | 1079 | |||
NM_001323981.1 | NP_001310910.1 | 6 | 1292 | |||
NM_194298.2 | NP_919274.1 | 5 | 1209 | |||
XM_017015883.1 | XP_016871372.1 | 5 | 1209 | |||
XM_017015884.1 | XP_016871373.1 | 6 | 1542 | |||
GenBank mRNA | AK125791.1 | - | 5 | 869 | ||
BC126238.1 | - | 4 | 798 | 8991954 | ||
BC130567.1 | - | 4 | 798 | 40146598 |