Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF542P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003127.2 | 7 | 1703 | |||
NR_024055.2 | 6 | 1606 | ||||
NR_024056.2 | 6 | 1859 | ||||
NR_024057.2 | 5 | 1787 | ||||
NR_033418.1 | 6 | 1927 | ||||
GenBank mRNA | AK097128.1 | - | 5 | 1591 | ||
AL833621.1 | - | 5 | 1556 | |||
BC089419.1 | - | 5 | 1531 | |||
BX640680.1 | - | 7 | 1663 | |||
BX647919.1 | - | 6 | 1662 |