Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287819.1 | NP_001274748.1 | 27 | 4934 | ||
NM_001287820.1 | NP_001274749.1 | 27 | 4805 | |||
NM_198503.3 | NP_940905.2 | 28 | 5006 | |||
XM_006711294.2 | XP_006711357.1 | 29 | 5501 | |||
XM_006711295.2 | XP_006711358.1 | 28 | 5429 | |||
XM_011509482.2 | XP_011507784.1 | 28 | 5426 | |||
XM_017001181.1 | XP_016856670.1 | 19 | 4016 | |||
XM_017001182.1 | XP_016856671.1 | 28 | 5392 | |||
XM_017001183.1 | XP_016856672.1 | 16 | 3552 | |||
XM_017001184.1 | XP_016856673.1 | 27 | 5371 | |||
GenBank mRNA | AK127807.1 | - | 20 | 4207 | ||
BX647852.1 | - | 27 | 4917 |