Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199069.1 | NP_951032.1 | 5 | 1026 | ||
NM_199070.1 | NP_951033.1 | 5 | 589 | |||
NM_199073.1 | NP_951047.1 | 5 | 660 | |||
NM_199074.1 | NP_951056.1 | 5 | 541 | |||
GenBank mRNA | BC002873.2 | - | 5 | 576 | 3945015 | |
BC111004.1 | - | 5 | 1656 | 5744973 | ||
BQ652614.1 | - | 5 | 541 | |||
CD358942.1 | - | 5 | 730 |