Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004467.3 | NP_004458.3 | 4, 5 | 523 | ||
NM_147203.2 | NP_671736.2 | 4, 5 | 611 | |||
NM_201552.1 | NP_963846.1 | 5, 6 | 650 | |||
NM_201553.1 | NP_963847.1 | 5, 6 | 738 | |||
XM_017013232.1 | XP_016868721.1 | 5, 6 | 1500 | |||
XM_017013233.1 | XP_016868722.1 | 3, 4 | 575 | |||
GenBank mRNA | AF042354.1 | - | 4, 5 | 559 | ||
AF168954.1 | - | 4, 5 | 546 | |||
AK225616.1 | - | 4, 5 | 509 | |||
AK225619.1 | - | 4, 5 | 498 | |||
AK289448.1 | - | 4, 5 | 509 | |||
AX772838.1 | - | 5, 6 | 603 | |||
BC007047.1 | - | 4, 5 | 519 | 4071810 | ||
BG567956.1 | - | 4, 5 | 618 | |||
BG618053.1 | - | 4, 5 | 524 | |||
BT006635.1 | - | 3, 4 | 414 | |||
D14446.1 | - | 4, 5 | 492 | |||
D87342.1 | - | 3, 4 | 419 |