Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004481.4 | NP_004472.1 | 1, 2 | 220 | ||
XM_017000963.1 | XP_016856452.1 | 1, 2 | 180 | |||
GenBank mRNA | AK290048.1 | - | 1, 2 | 146 | ||
AK296886.1 | - | 1, 2 | 177 | |||
BC041120.1 | - | 1, 2 | 206 | 5553465 | ||
BG773173.1 | - | 1, 2 | 225 | |||
X85019.1 | - | 1, 2 | 134 |