Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297709.1 | NP_001284638.1 | 3, 4 | 360 | ||
NM_001297711.1 | NP_001284640.1 | 3, 4 | 360 | |||
NM_001297712.1 | NP_001284641.1 | 3, 4 | 360 | |||
NM_003480.3 | NP_003471.1 | 3, 4 | 360 | |||
NR_123733.1 | 3, 4 | 360 | ||||
NR_123734.1 | 3, 4 | 360 | ||||
GenBank mRNA | AK124368.1 | - | 3, 4 | 356 | ||
AK290585.1 | - | 3, 4 | 151 | |||
AK299475.1 | - | 3, 4 | 188 | |||
AK315807.1 | - | 3, 4 | 147 | |||
AU280458.1 | - | 3, 4 | 298 | |||
AY339060.1 | - | 2, 3 | 96 | |||
BC005901.1 | - | 3, 4 | 150 | 4247343 | ||
BU929788.1 | - | 3, 4 | 164 | |||
DA544968.1 | - | 3, 4 | 197 | |||
DB247952.1 | - | 3, 4 | 360 | |||
DN996275.1 | - | 3, 4 | 181 | |||
U37283.1 | - | 3, 4 | 282 |