Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261834.1 | NP_001248763.1 | 2 | 306 | ||
NM_004749.3 | NP_004740.2 | 2 | 350 | |||
NM_030900.3 | NP_112162.1 | 2 | 350 | |||
NM_199122.2 | NP_954573.1 | 2 | 329 | |||
GenBank mRNA | AB023165.1 | - | 2 | 321 | ||
AK001281.1 | - | 2 | 298 | |||
AK022575.1 | - | 2 | 286 | |||
AK222943.1 | - | 2 | 286 | |||
AK300489.1 | - | 2 | 265 | |||
AL833840.1 | - | 2 | 319 | |||
BC014918.1 | - | 2 | 280 | 3906670 | ||
BC017235.2 | - | 2 | 277 | 3538504 | ||
BG703429.1 | - | 2 | 315 | |||
DA980367.1 | - | 2 | 350 |