Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADIPOQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177800.1 | NP_001171271.1 | 4 | 1242 | ||
NM_004797.3 | NP_004788.1 | 3 | 1191 | |||
GenBank mRNA | AX767959.1 | - | 3 | 1133 | ||
D45371.1 | - | 3 | 1133 |