Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164617.1 | NP_001158089.1 | 9 | 2076 | ||
NM_001164618.1 | NP_001158090.1 | 8 | 1959 | |||
NM_001164619.1 | NP_001158091.1 | 7 | 1845 | |||
NM_004484.3 | NP_004475.1 | 8 | 2007 | |||
GenBank mRNA | AK222761.1 | - | 8 | 1955 | ||
AK222766.1 | - | 8 | 1955 | |||
AK300168.1 | - | 8 | 1930 | |||
BC035972.1 | - | 8 | 1955 | 4603748 | ||
L47125.1 | - | 8 | 1961 | |||
L47176.1 | - | 8 | 1956 | |||
U50410.1 | - | 8 | 1954 | |||
Z37987.1 | - | 8 | 1918 |