Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286132.1 | NP_001273061.1 | 5 | 635 | ||
NM_001286133.1 | NP_001273062.1 | 5 | 578 | |||
NM_006638.3 | NP_006629.2 | 6 | 704 | |||
XM_011514253.2 | XP_011512555.1 | 6 | 699 | |||
GenBank mRNA | BC017871.1 | - | 5 | 609 | 4278825 | |
BQ220155.1 | - | 6 | 652 |