Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007001.2 | NP_008932.2 | 7 | 637 | ||
NR_104627.1 | 7 | 637 | ||||
XM_005251673.1 | XP_005251730.1 | 7 | 612 | |||
XM_005251674.1 | XP_005251731.1 | 7 | 612 | |||
XM_005251675.2 | XP_005251732.1 | 5 | 481 | |||
XM_005251676.1 | XP_005251733.1 | 7 | 612 | |||
XM_005251678.4 | XP_005251735.1 | 4 | 276 | |||
GenBank mRNA | AB106537.1 | - | 7 | 570 | ||
AB122077.1 | - | 7 | 582 | |||
AJ005866.1 | - | 5 | 333 | |||
AK307890.1 | - | 7 | 617 | |||
BC009413.1 | - | 1 | 28 | |||
BC113579.1 | - | 8 | 590 | 8322631 | ||
BX361048.2 | - | 7 | 636 |