Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007001.2 | NP_008932.2 | 8, 9 | 768 | ||
NR_104627.1 | 8, 9 | 768 | ||||
XM_005251674.1 | XP_005251731.1 | 8, 9 | 743 | |||
XM_005251675.2 | XP_005251732.1 | 6, 7 | 612 | |||
XM_005251678.4 | XP_005251735.1 | 5, 6 | 407 | |||
XM_011518164.1 | XP_011516466.1 | 6, 7 | 582 | |||
GenBank mRNA | AB106537.1 | - | 8, 9 | 701 | ||
AB122077.1 | - | 8, 9 | 713 | |||
AJ005866.1 | - | 6, 7 | 464 | |||
AK307890.1 | - | 8, 9 | 748 | |||
BC009413.1 | - | 2, 3 | 159 | |||
BC113579.1 | - | 9, 10 | 721 | 8322631 | ||
BX361048.2 | - | 8, 9 | 768 |