Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301016.2 | NP_001287945.1 | 14, 15 | 2449 | ||
NM_001321208.1 | NP_001308137.1 | 14, 15 | 2489 | |||
NM_001321211.1 | NP_001308140.1 | 14, 15 | 2273 | |||
NM_007040.5 | NP_008971.2 | 14, 15 | 2780 | |||
NM_144732.4 | NP_653333.1 | 14, 15 | 2262 | |||
XM_005258459.3 | XP_005258516.1 | 14, 15 | 2613 | |||
XM_005258461.4 | XP_005258518.1 | 14, 15 | 2443 | |||
XM_005258463.3 | XP_005258520.1 | 14, 15 | 2434 | |||
XM_005258464.3 | XP_005258521.1 | 14, 15 | 2249 | |||
XM_005258465.4 | XP_005258522.1 | 14, 15 | 2460 | |||
XM_011526393.2 | XP_011524695.1 | 14, 15 | 2535 | |||
XM_011526394.2 | XP_011524696.1 | 14, 15 | 2290 | |||
XM_011526395.2 | XP_011524697.1 | 14, 15 | 2499 | |||
GenBank mRNA | AA479761.1 | - | 1, 2 | 79 | ||
AJ007509.1 | - | 14, 15 | 2623 | |||
AK001229.1 | - | 5, 6 | 1046 | |||
AK021455.1 | - | 3, 4 | 1065 | |||
AK022863.1 | - | 14, 15 | 2582 | |||
AK023006.1 | - | 14, 15 | 2219 | |||
AK127057.1 | - | 13, 14 | 2270 | |||
AK225850.1 | - | 14, 15 | 2272 | |||
AK290729.1 | - | 14, 15 | 2244 | |||
AK291315.1 | - | 15, 16 | 2529 | |||
AK291762.1 | - | 14, 15 | 2582 | |||
AK308156.1 | - | 15, 16 | 2292 | |||
AL050146.1 | - | 7, 8 | 1388 | |||
BC002564.2 | - | 14, 15 | 2520 | 3139049 | ||
BC004242.1 | - | 2, 3 | 424 | |||
BC009988.2 | - | 14, 15 | 2426 | 4127873 | ||
BC014232.2 | - | 14, 15 | 2401 | 4558792 | ||
BC027713.2 | - | 14, 15 | 2347 | 4123077 |