Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301016.2 | NP_001287945.1 | 15 | 3271 | ||
NM_001321208.1 | NP_001308137.1 | 15 | 3311 | |||
NM_001321211.1 | NP_001308140.1 | 15 | 3095 | |||
NM_007040.5 | NP_008971.2 | 15 | 3602 | |||
NM_144732.4 | NP_653333.1 | 15 | 3084 | |||
XM_005258459.3 | XP_005258516.1 | 15 | 3435 | |||
XM_005258461.4 | XP_005258518.1 | 15 | 3265 | |||
XM_005258463.3 | XP_005258520.1 | 15 | 3256 | |||
XM_005258464.3 | XP_005258521.1 | 15 | 3071 | |||
XM_005258465.4 | XP_005258522.1 | 15 | 3282 | |||
XM_011526392.2 | XP_011524694.1 | 14 | 3213 | |||
XM_011526393.2 | XP_011524695.1 | 15 | 3357 | |||
XM_011526394.2 | XP_011524696.1 | 15 | 3112 | |||
XM_011526395.2 | XP_011524697.1 | 15 | 3321 | |||
GenBank mRNA | AK022863.1 | - | 15 | 3404 | ||
AK225850.1 | - | 15 | 3094 | |||
AL050146.1 | - | 8 | 2210 | |||
BC000860.1 | - | 1 | 415 | |||
BM976139.1 | - | 1 | 36 |