Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012416.3 | NP_036548.1 | 1 | 563 | ||
GenBank mRNA | AF039023.1 | - | 1 | 532 | ||
AK307840.1 | - | 1 | 538 | |||
BC098406.1 | - | 1 | 538 | 30347107 | ||
BX537405.1 | - | 1 | 542 |