Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243202.1 | NP_001230131.1 | 2 | 342 | ||
NM_001243203.1 | NP_001230132.1 | 2 | 283 | |||
NM_012416.3 | NP_036548.1 | 1 | 1398 | |||
XM_017014624.1 | XP_016870113.1 | 2 | 303 | |||
GenBank mRNA | AF039023.1 | - | 1 | 1367 | ||
AK300408.1 | - | 2 | 257 | |||
AK307840.1 | - | 1 | 1373 | |||
BC098406.1 | - | 1 | 1373 | 30347107 | ||
BX537405.1 | - | 1 | 1377 | |||
DA234653.1 | - | 2 | 327 | |||
DC342914.1 | - | 2 | 267 |