Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243202.1 | NP_001230131.1 | 2 | 3430 | ||
NM_001243203.1 | NP_001230132.1 | 2 | 3371 | |||
NM_012416.3 | NP_036548.1 | 1 | 4486 | |||
XM_017014624.1 | XP_016870113.1 | 2 | 3391 | |||
GenBank mRNA | AK001982.1 | - | 1 | 1817 | ||
BC012805.2 | - | 1 | 2426 | |||
BC098406.1 | - | 1 | 4460 | 30347107 | ||
BX537405.1 | - | 1 | 4465 |