Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280796.1 | NP_001267725.1 | 2 | 556 | ||
NM_001280797.1 | NP_001267726.1 | 2 | 556 | |||
NM_013267.3 | NP_037399.2 | 2 | 556 | |||
XM_005268797.1 | XP_005268854.1 | 2 | 183 | |||
XM_017019180.1 | XP_016874669.1 | 2 | 183 | |||
GenBank mRNA | AF223944.1 | - | 2 | 483 | ||
AK000467.1 | - | 2 | 303 | |||
AK289471.1 | - | 2 | 260 | |||
AK294822.1 | - | 2 | 501 | |||
AK303772.1 | - | 2 | 391 | |||
BC025661.1 | - | 2 | 271 | |||
BC119767.1 | - | 2 | 501 | |||
BC119768.1 | - | 2 | 501 | |||
BI916699.1 | - | 2 | 543 | |||
HM775422.1 | - | 1 | 175 | |||
HM775423.1 | - | 1 | 175 | |||
HM775424.1 | - | 1 | 175 |