Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014023.3 | NP_054742.2 | 12, 13 | 1419 | ||
GenBank mRNA | AB023199.1 | - | 12, 13 | 1390 | ||
AK128194.1 | - | 11, 12 | 4205 | |||
AK292591.1 | - | 12, 13 | 1385 | |||
AK307259.1 | - | 11, 12 | 1312 | |||
AL136827.1 | - | 12, 13 | 1397 |