Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126050.1 | NP_001119522.1 | 6 | 1695 | ||
NM_001126051.1 | NP_001119523.1 | 6 | 1670 | |||
NM_001319186.1 | NP_001306115.1 | 6 | 1716 | |||
NM_001319187.1 | NP_001306116.1 | 6 | 1520 | |||
NM_001319188.1 | NP_001306117.1 | 7 | 1685 | |||
NM_004494.2 | NP_004485.1 | 6 | 1884 | |||
NR_135008.1 | 7 | 1765 | ||||
XM_011509453.2 | XP_011507755.2 | 6 | 1707 | |||
XM_011509454.2 | XP_011507756.1 | 6 | 1537 | |||
XM_011509455.2 | XP_011507757.1 | 6 | 1603 | |||
GenBank mRNA | AK096411.1 | - | 6 | 1671 | ||
AL833592.1 | - | 8 | 1977 | |||
BC018991.1 | - | 7 | 1577 | 3503321 | ||
D16431.1 | - | 6 | 1876 |