Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014109.3 | NP_054828.2 | 7 | 992 | ||
XM_011516994.2 | XP_011515296.1 | 7 | 1519 | |||
XM_011516995.2 | XP_011515297.1 | 7 | 1519 | |||
GenBank mRNA | AK097133.1 | - | 7 | 974 | ||
AL833653.1 | - | 7 | 746 | |||
AY598335.1 | - | 7 | 884 | |||
BC113656.1 | - | 7 | 906 | 8322708 | ||
CR749832.1 | - | 8 | 1248 |