Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282708.1 | NP_001269637.1 | 8 | 1547 | ||
NM_001282709.1 | NP_001269638.1 | 8 | 1535 | |||
NM_014285.6 | NP_055100.2 | 9 | 1625 | |||
NR_104230.1 | 8 | 2131 | ||||
XM_005272176.2 | XP_005272233.1 | 9 | 1623 | |||
XM_006717023.2 | XP_006717086.1 | 8 | 1533 | |||
XM_017014558.1 | XP_016870047.1 | 8 | 2157 | |||
GenBank mRNA | AK001916.1 | - | 9 | 1614 | ||
AK022460.1 | - | 9 | 1611 | |||
AK090512.1 | - | 8 | 2157 | |||
AK092727.1 | - | 3 | 2391 | |||
BC000747.2 | - | 9 | 1595 | 2821702 |