Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014640.4 | NP_055455.3 | 12 | 2804 | ||
XM_006712875.2 | XP_006712938.1 | 12 | 2814 | |||
XM_017005387.1 | XP_016860876.1 | 12 | 2814 | |||
XM_017005388.1 | XP_016860877.1 | 13 | 2701 | |||
XM_017005389.1 | XP_016860878.1 | 13 | 2720 | |||
XM_017005390.1 | XP_016860879.1 | 12 | 2654 | |||
XM_017005391.1 | XP_016860880.1 | 11 | 2719 | |||
XM_017005392.1 | XP_016860881.1 | 10 | 2969 | |||
GenBank mRNA | AK091026.1 | - | 2 | 1927 | ||
AK291770.1 | - | 12 | 2762 | |||
AK296054.1 | - | 5 | 554 | |||
AK302728.1 | - | 12 | 2350 | |||
AL833511.1 | - | 12 | 2453 | |||
BC021707.2 | - | 12 | 2729 | 4824604 | ||
D79995.1 | - | 12 | 2641 |