Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023570.3 | NP_001018864.2 | 15 | 2028 | ||
NM_001023571.3 | NP_001018865.2 | 12 | 1629 | |||
NM_001319107.1 | NP_001306036.1 | 15 | 2027 | |||
NR_134968.1 | 14 | 1917 | ||||
XM_005247911.3 | XP_005247968.1 | 14 | 1870 | |||
XM_005247912.2 | XP_005247969.1 | 14 | 1841 | |||
XM_011513335.2 | XP_011511637.1 | 13 | 1747 | |||
XM_017007537.1 | XP_016863026.1 | 14 | 1910 | |||
XM_017007538.1 | XP_016863027.1 | 11 | 1712 | |||
XM_017007539.1 | XP_016863028.1 | 11 | 1458 | |||
GenBank mRNA | AB062481.1 | - | 14 | 1911 | ||
AY714228.1 | - | 15 | 2022 | |||
AY964667.1 | - | 12 | 1579 | |||
BC005806.2 | - | 12 | 1570 | 3834623 | ||
D25278.1 | - | 15 | 1969 |