Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286184.1 | NP_001273113.1 | 4 | 2547 | ||
NM_014655.3 | NP_055470.1 | 4 | 2523 | |||
NR_104408.1 | 4 | 2046 | ||||
NR_104411.1 | 3 | 1885 | ||||
NR_104412.1 | 2 | 1757 | ||||
XM_006711657.3 | XP_006711720.1 | 5 | 2929 | |||
XM_011510180.1 | XP_011508482.1 | 6 | 3280 | |||
XM_011510181.1 | XP_011508483.1 | 5 | 3105 | |||
XM_017002904.1 | XP_016858393.1 | 6 | 3256 | |||
XM_017002905.1 | XP_016858394.1 | 5 | 3081 | |||
XM_017002906.1 | XP_016858395.1 | 5 | 2902 | |||
XM_017002907.1 | XP_016858396.1 | 3 | 2243 | |||
XM_017002908.1 | XP_016858397.1 | 4 | 2953 | |||
GenBank mRNA | AB007915.3 | - | 4 | 2346 | ||
AK074912.1 | - | 4 | 2372 | |||
AK075002.1 | - | 4 | 2347 | |||
BC008843.2 | - | 4 | 2349 | 4136523 | ||
BC039854.1 | - | 4 | 2334 | 6061399 |