Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242795.1 | NP_001229724.1 | 17 | 2224 | ||
NM_001242796.1 | NP_001229725.1 | 17 | 2173 | |||
NM_014669.4 | NP_055484.3 | 19 | 2219 | |||
XM_005256263.3 | XP_005256320.1 | 19 | 2155 | |||
GenBank mRNA | AK056637.1 | - | 17 | 2187 | ||
AK292262.1 | - | 18 | 2127 | |||
AK294176.1 | - | 17 | 2218 | |||
BC034346.1 | - | 19 | 2129 | 4750923 | ||
D42085.1 | - | 19 | 2164 |