Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL11RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142784.2 | NP_001136256.1 | 9 | 891 | ||
NR_052010.1 | 9 | 945 | ||||
GenBank mRNA | AK075530.1 | - | 9 | 942 | ||
AK094689.1 | - | 9 | 2198 | |||
AK292073.1 | - | 9 | 925 | |||
BC003110.1 | - | 9 | 907 | 3502059 | ||
BT009864.1 | - | 8 | 842 | |||
U32324.1 | - | 9 | 903 | |||
Z38102.1 | - | 8 | 847 | |||
Z46595.1 | - | 8 | 836 |