Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014798.2 | NP_055613.1 | 3 | 343 | ||
NR_027782.1 | 3 | 343 | ||||
XM_011525523.2 | XP_011523825.1 | 3 | 314 | |||
XM_011525524.1 | XP_011523826.1 | 3 | 493 | |||
XM_011525526.1 | XP_011523828.1 | 3 | 338 | |||
XM_017025451.1 | XP_016880940.1 | 3 | 291 | |||
XM_017025453.1 | XP_016880942.1 | 3 | 314 | |||
XM_017025454.1 | XP_016880943.1 | 3 | 314 | |||
GenBank mRNA | AB002354.2 | - | 3 | 296 | ||
BC064361.1 | - | 3 | 307 | 5535600 | ||
DB064851.1 | - | 3 | 377 |
Gene Symbol: PLEKHM1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK304173.1 | - | 3 | 284 | ||
AK308180.1 | - | 3 | 307 | |||
BC034693.1 | - | 3 | 295 | |||
DA759804.1 | - | 3 | 344 |