Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014798.2 | NP_055613.1 | 5 | 1129 | ||
NR_027774.1 | 4 | 881 | ||||
XM_006722201.3 | XP_006722264.1 | 3 | 910 | |||
XM_011525523.2 | XP_011523825.1 | 5 | 1100 | |||
XM_011525524.1 | XP_011523826.1 | 5 | 1279 | |||
XM_011525525.1 | XP_011523827.1 | 4 | 887 | |||
XM_011525526.1 | XP_011523828.1 | 6 | 923 | |||
XM_017025451.1 | XP_016880940.1 | 5 | 1077 | |||
XM_017025452.1 | XP_016880941.1 | 4 | 832 | |||
XM_017025453.1 | XP_016880942.1 | 6 | 1286 | |||
GenBank mRNA | AB002354.2 | - | 5 | 1082 | ||
AJ002220.1 | - | 3 | 717 | |||
AK299116.1 | - | 4 | 829 | |||
AK299467.1 | - | 4 | 883 | |||
BC064361.1 | - | 5 | 1093 | 5535600 |