Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014798.2 | NP_055613.1 | 12 | 4436 | ||
NR_027774.1 | 11 | 4188 | ||||
NR_027782.1 | 11 | 4051 | ||||
XM_006722201.3 | XP_006722264.1 | 10 | 4217 | |||
XM_011525524.1 | XP_011523826.1 | 12 | 4586 | |||
XM_011525525.1 | XP_011523827.1 | 11 | 4194 | |||
XM_011525526.1 | XP_011523828.1 | 13 | 4230 | |||
XM_011525528.2 | XP_011523830.1 | 7 | 2780 | |||
XM_017025451.1 | XP_016880940.1 | 12 | 4384 | |||
XM_017025452.1 | XP_016880941.1 | 11 | 4139 | |||
XM_017025453.1 | XP_016880942.1 | 13 | 4593 | |||
XM_017025454.1 | XP_016880943.1 | 11 | 4284 | |||
GenBank mRNA | AB002354.2 | - | 12 | 4389 | ||
AK074103.1 | - | 8 | 3184 | |||
AK299116.1 | - | 11 | 4136 | |||
BC064361.1 | - | 12 | 4400 | 5535600 |