Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079673.1 | NP_001073141.1 | 5 | 614 | ||
NM_001278438.1 | NP_001265367.1 | 5 | 657 | |||
NM_014923.4 | NP_055738.3 | 3 | 238 | |||
NR_103528.1 | 5 | 657 | ||||
XM_011534997.2 | XP_011533299.1 | 4 | 328 | |||
XM_017020440.1 | XP_016875929.1 | 5 | 499 | |||
XM_017020441.1 | XP_016875930.1 | 7 | 1120 | |||
XM_017020442.1 | XP_016875931.1 | 6 | 652 | |||
XM_017020443.1 | XP_016875932.1 | 5 | 972 | |||
GenBank mRNA | AB023187.2 | - | 3 | 168 | ||
AJ749706.1 | - | 5 | 616 | |||
AJ749707.1 | - | 3 | 229 | |||
AK056071.1 | - | 5 | 598 | |||
AK302415.1 | - | 3 | 238 | |||
BC036010.1 | - | 1 | 34 | |||
BC107793.1 | - | 3 | 159 | |||
BC132812.1 | - | 5 | 381 | 40146834 | ||
BC136617.1 | - | 5 | 382 | 9020607 | ||
BC144301.1 | - | 5 | 382 | 9052821 | ||
BX640739.1 | - | 5 | 517 | |||
BX648141.1 | - | 5 | 604 | |||
BX648706.1 | - | 5 | 612 | |||
DB460317.2 | - | 5 | 657 |