Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014975.2 | NP_055790.1 | 2 | 228 | ||
XM_011527805.2 | XP_011526107.1 | 3 | 125 | |||
GenBank mRNA | AB023190.1 | - | 2 | 139 | ||
AK297553.1 | - | 2 | 132 | |||
DA769770.1 | - | 2 | 228 |