Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004522.2 | NP_004513.1 | 4, 5 | 799 | ||
XM_017004062.1 | XP_016859551.1 | 4, 5 | 790 | |||
GenBank mRNA | AB011103.1 | - | 4, 5 | 772 | ||
BC002721.2 | - | 4, 5 | 781 | |||
BC017298.2 | - | 4, 5 | 754 | |||
BC025961.1 | - | 4, 5 | 781 | |||
BC127772.1 | - | 1, 2 | 123 |