Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318328.1 | NP_001305257.1 | 2 | 532 | ||
NM_015045.3 | NP_055860.1 | 2 | 532 | |||
XM_011539548.1 | XP_011537850.1 | 2 | 564 | |||
XM_017015979.1 | XP_016871468.1 | 2 | 564 | |||
XM_017015980.1 | XP_016871469.1 | 2 | 525 | |||
GenBank mRNA | AB065003.1 | - | 2 | 535 | ||
AB426821.1 | - | 2 | 487 | |||
AB426822.1 | - | 4 | 437 | |||
AB426823.1 | - | 3 | 430 | |||
AB426824.1 | - | 3 | 354 | |||
AB426825.1 | - | 3 | 328 | |||
AB426826.1 | - | 3 | 317 | |||
AB426827.1 | - | 3 | 297 | |||
AB426828.1 | - | 3 | 249 | |||
AB426829.1 | - | 2 | 245 | |||
AB426830.1 | - | 2 | 239 | |||
AB426831.1 | - | 3 | 228 | |||
AB426832.1 | - | 2 | 226 | |||
AB426833.1 | - | 3 | 222 | |||
AB426834.1 | - | 3 | 210 | |||
AB426835.1 | - | 3 | 180 | |||
AB426836.1 | - | 2 | 153 | |||
AF479418.1 | - | 3 | 228 | |||
AK290138.1 | - | 2 | 493 | |||
BC140858.1 | - | 2 | 256 | 9021726 | ||
BC150269.1 | - | 3 | 383 | 9007232 | ||
D87450.1 | - | 3 | 383 |