Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318328.1 | NP_001305257.1 | 2 | 723 | ||
NM_015045.3 | NP_055860.1 | 2 | 723 | |||
XM_011539548.1 | XP_011537850.1 | 2 | 755 | |||
XM_017015979.1 | XP_016871468.1 | 2 | 755 | |||
XM_017015980.1 | XP_016871469.1 | 2 | 716 | |||
GenBank mRNA | AB065003.1 | - | 2 | 726 | ||
AF479418.1 | - | 3 | 419 | |||
AK290138.1 | - | 2 | 684 | |||
BC140858.1 | - | 2 | 447 | 9021726 | ||
BC150269.1 | - | 3 | 574 | 9007232 | ||
D87450.1 | - | 3 | 574 |